Learn More
STEMCELL Technologies Anti-Rhesus Red Blood Cell Antibody, Clone T4G6, Size: 100 µg
SDP
Supplier:  STEMCELL Technologies 60133
Encompass
The mouse monoclonal antibody T4G6 recognizes mature red blood cells and erythroid precursor cells in rhesus monkey bone marrow. Antibody clone T4G6 has been developed in mice immunized with rhesus monkey (Macaca mulatta) red blood cells. The antigen to which clone T4G6 binds has not been identified. The reactivity with red blood cells from other monkey species has not been determined. Clone T4G6 does not recognize human red blood cells.
Catalog No. 50-197-5705