Learn More
AdipoGen anti-RIG-I, mAb (Alme-1),Monoclonal Antibody.
SDP
Supplier:  AdipoGen AG20B0009C100
Encompass_Preferred
anti-RIG-I, mAb (Alme-1), AdipoGen Life Sciences. Monoclonal Antibody. Recognizes human and mouse RIG-I. Isotype: Mouse IgG1. Clone: Alme-1. Applications: IHC, IP, WB. Liquid. In PBS containing 10% glycerol and 0.02% sodium azide. RIG-I and MDA5 are highly conserved helicases involved in the innate immune response to virus. RIG-I is a member of the DEAD-box RNA helicases and is activated by cytoplasmic dsRNA and 5¿-ppp RNA produced during the viral replication. The protein is characterized by a N-terminal region with two caspase recruitment domains (CARD) and a C-terminal region harboring potential ATP-dependent RNA helicase activity. RIG-I recruits the CARD adaptor inducing IFN-beta (Cardif) in a CARD-CARD-dependent manner resulting in NF-kappaB and IRF3 activation.
Catalog No. 50-463-373