Learn More
ACROBiosystems ACROBiosystems Anti-Rituximab Antibodies (MALS verified)
Supplier: ACROBiosystems RIBY35C100UG
Antibody;100UG;IgG1 / kappa;Recognizes Rituximab specifically, no cross reactivity with other humanized antibodies.;Anti-Rituximab Antibodies (RIB-Y35c) are expressed from human 293 cells (HEK293).;This protein . ;Immobilized Rituximab at 1 μg/mL (100 μL/well) can bind Anti-Rituximab Antibodies (Cat. No. RIB-Y35c) with a linear range of 0.2-6 ng/mL (QC tested).