Learn More
LAB BIOREAGENTS Anti-Rituximab Elisa Kit
Catalog No. 501959292
Encompass
Anti-Rituximab ELISA Kit is a sandwich enzyme-linked immunoassay for the quantitative determination of Anti-Rituximab with high sensitivity and specificity in serum and plasma.Kit PerformanceSensitivity: 40 ng/mLCross-reactivity: There is no cross reaction with native serum immunoglobulinsPrecision: Intra-Assay: CV < 20%; Inter-Assay: CV < 20%
Catalog No. 50-195-9292 Supplier LAB BIOREAGENTS Supplier No. REK021215
Anti-Rituximab ELISA Kit is a sandwich enzyme-linked immunoassay for the quantitative determination of Anti-Rituximab with high sensitivity and specificity in serum and plasma.Kit PerformanceSensitivity: 40 ng/mLCross-reactivity: There is no cross reaction with native serum immunoglobulinsPrecision: Intra-Assay: CV < 20%; Inter-Assay: CV < 20%