Learn More
KRISHGEN BIOSYSTEMS Immunoassay for the Quantitative Determination of Anti-Rituximab antibodies in human serum and plasma. Assay Range: 10 - 640 ng/ml; Sensitivity: 10 ng/ml.
Supplier:  KRISHGEN BIOSYSTEMS KBI2010
Encompass_Preferred
Pack Size: 1 x 96 wells. This ELISA is optimised to detect antibodies to rituximab in human samples. If you would like to use animal samples, please contact us. Introduction: Rituximab is a monoclonal antibody which has high specificity for CD20 and it is used to treat certain autoimmune diseases and types of cancer. It is also used for non-Hodgkin lymphoma, chronic lymphocytic leukemia, rheumatoid arthritis, and granulomatosis with polyangiitis, idiopathic thrombocytopenic purpura, pemphigus vulgaris, myasthenia gravis and Epstein - Barr virus-positive mucocutaneous ulcers. It is given by slow injection into a vein. Anti-Drug Antibodies (ADA) may induce unwanted side effects in biopharmaceuticals. Hence, ADA has been subjected to increase in scrutiny by the regulatory authorities using immunogenicity safety studies. ELISA Features include a pre-coated plate with break-apart wells, standardisation and Precision CV <10%.
Catalog No. NC2747148