Learn More
Leica Biosystems Anti-roll plate, glass 50 mm for knife holder CN
Catalog No. NC0458774
Encompass
Anti-roll plate glass 50 mm is a glass insert for cryostat anti-roll assemblies that helps prevent tissue sections from curling during sectioning and provides a stable surface for guiding sections after cutting.
  • Prevents tissue section roll-up during cryostat sectioning.
  • 50 mm glass insert for anti-roll assemblies.
  • Provides a smooth, stable surface for consistent section collection.
  • Durable glass construction for reliable performance.
  • Compatible with common cryostat anti-roll holders and knife guide assemblies.
Catalog No. NC0458774 Supplier Leica Biosystems Supplier No. 14041933816
Anti-roll plate glass 50 mm is a glass insert for cryostat anti-roll assemblies that helps prevent tissue sections from curling during sectioning and provides a stable surface for guiding sections after cutting.
  • Prevents tissue section roll-up during cryostat sectioning.
  • 50 mm glass insert for anti-roll assemblies.
  • Provides a smooth, stable surface for consistent section collection.
  • Durable glass construction for reliable performance.
  • Compatible with common cryostat anti-roll holders and knife guide assemblies.