Learn More
Aviva Systems Biology Corp RP11-269F19.9 Antibody - N-terminal region (ARP54425_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54425_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RP11-269F19.9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RP11-269F19.9. Alternate names: LOC343521, RP11-269F19.9ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53836