Learn More
Aviva Systems Biology Corp RP11-298P3.3 Antibody - N-terminal region (ARP55406_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55406_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RP11-298P3.3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RP11-298P3.3. Alternate names: 92M18.3, CG005, FLJ36195, FLJ41089, FLJ43077, PFAAP5, RP11-298P3.3ApplicationWB. Species reactivity: Dog, Horse, Human, Pig
Catalog No. 50-014-53842