Learn More
Aviva Systems Biology Corp RP11-50D16.3 Antibody - middle region (ARP44638_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44638_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RP11-50D16.3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RP11-50D16.3. Alternate names: DKFZp313M1221, DKFZp686E1140, LOC387921, RP11-50D16.3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53829