Learn More
Aviva Systems Biology Corp RP11-529I10.4 Antibody - middle region (ARP55257_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55257_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RP11-529I10.4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RP11-529I10.4. Alternate names: DKFZP566F084, RP11-529I10.4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53838