Learn More
Aviva Systems Biology Corp HNRNPA1L2 Antibody - N-terminal region (ARP40282_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40282_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RP11-78J21.1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human HNRNPA1L2.. Alternate names: ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53827