Learn More
Aviva Systems Biology Corp KYAT3 Antibody - N-terminal region (ARP40267_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40267_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RP11-82K18.3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RP11-82K18.3. Alternate names: RP11-82K18.3, RP4-531M19.2, KAT3, KATIIIApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53826