Learn More
Aviva Systems Biology Corp ARHGAP36 Antibody - N-terminal region (ARP52766_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52766_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RP13-102H20.1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human RP13-102H20.1. Alternate names: FLJ30058, RP13-102H20.1ApplicationWB. Species reactivity: Goat, Horse, Human, Mouse, Pig, Rabbit, Sheep
Catalog No. 50-014-53834