Learn More
Description
- Made from Heat-sealable material where each bag emits an odorless, invisible, and non-toxic vapor that actively prevents oxidation on ferrous metal surfaces.
- Protects weapons during Long-term storage
- Vapors emitted does not stick to or damage weapons
- Features a Heavy-duty construction.
- Designed to be heat sealable.
- Features an Anti-rust design.
- 4 mm thick polybag
- Available in 7 sizes
Specifications
Specifications
| Dimensions (L x W) | 9 x 12 in. |
| Product Type | Evidence Bag |
| Color | Transparent with Yellow Back |
| Quantity | 100/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
