Learn More
Sino Biological Anti-S100A9 Antibody, Rabbit Polyclonal 100 µL
Catalog No. 5000750340
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human S100A9 (rh S100A9; Catalog#11145-H08B; NP_002956.1; Met1-Pro114). S100A9 specific IgG was purified by Human S100A9 affinity chromatography.
Catalog No. 50-007-50340 Supplier Sino Biological Supplier No. 11145T62100
Produced in rabbits immunized with purified, recombinant Human S100A9 (rh S100A9; Catalog#11145-H08B; NP_002956.1; Met1-Pro114). S100A9 specific IgG was purified by Human S100A9 affinity chromatography.