Learn More
Seracare Life Sciences Inc ANTI-SALMONELLA CSA-1 BIOTINA
Supplier:  Seracare Life Sciences Inc 53600031
Encompass_Preferred
Affinity purified antibody isolated from a pool of serum from goats immunized with Salmonella species was labeled with a biotin analog having a long carbon spacer arm. This minimizes steric hindrance and maximizes binding.
Catalog No. 50-672-94