Learn More
Genscript Corporation Antibody Antibody Antibodies Antibodies 40 UG 1/EA
Supplier:  Genscript Corporation A0204340
Encompass
The product is specific for Sarilumab. The antibody is recommended as a detection antibody in a pharmacokinetic (PK) bridging assay with capture antibody GenScript A02042-40 Anti-Sarilumab Antibody (26B10) mAb Mouse
Catalog No. 50-253-0056