Learn More
ACROBiosystems Anti-SARS-CoV-2 Nucleocapsid Antibody, Mouse IgG1 (AS47) (Trehalose free)
Supplier:  ACROBiosystems NUNS47100MG
Encompass_Preferred
Antibody;100MG;Mouse IgG1/kappa;This product is a specific antibody against SARS-CoV-2 Nucleocapsid protein. No cross-reactivity is deteced with nucleocapsid protein of other coronaviruses, including SARS-CoV, MERS-CoV, HCoV-229E, HCoV-NL63, HCoV-OC43 and HCoV-HKU1.;Anti-SARS-CoV-2 Nucleocapsid Antibody, Mouse IgG1 (NUN-S47) was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified recombinant SARS-CoV-2 Nucleocapsid protein. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.; the protein migrates as 25 kDa and 55 kDa under reducing (R) condition, and 130 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized SARS-CoV-2 Nucleocapsid protein, His Tag (Cat. No. NUN-C5227) at 2 μg/mL (100 μL/well) can bind Anti-SARS-CoV-2 Nucleocapsid Antibody, Mouse IgG1 (Cat. No. NUN-S47) with a linear range of 0.15-2.5 ng/mL (QC tested).;Detection SARS-CoV-2 Nuc
Catalog No. 50-205-5260
Add to Cart
Request a Quote