Learn More
ACROBiosystems ACROBiosystems Anti-SARS-CoV-2 Nucleocapsid Antibody, Mouse IgG1 (AS47) (Trehalose free)
Supplier:  ACROBiosystems NUNS471.5MG
Encompass_Preferred
Antibody;1.5MG;Mouse IgG1/kappa;This product is a specific antibody against SARS-CoV-2 Nucleocapsid protein. No cross-reactivity is detected with nucleocapsid protein of other coronaviruses, including SARS-CoV, MERS-CoV, HCoV-229E, HCoV-NL63, HCoV-OC43 and HCoV-HKU1.;Anti-SARS-CoV-2 Nucleocapsid Antibody, Mouse IgG1 (NUN-S47) was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified recombinant SARS-CoV-2 Nucleocapsid protein. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography. ELISA test validated that this antibody can bind multiple N protein variants (Cat. No. NUN-C52H8, NUN-C52Hc, NUN-C52Hd, NUN-C52Hm, NUN-C52H6, NUN-C52H3, NUN-C52H9, NUN-C52H4) with similar affinity as compared to the wild type N protein (Cat. No. NUN-C5227).; the protein migrates as 25 kDa and 55 kDa under reducing (R) condition, and 130 kDa under non-reducing (NR) condition (SDS-PAGE).;I
Catalog No. 50-210-7460