Learn More
ACROBiosystems Anti-SARS-CoV-2 Spike RBD Antibody, Chimeric mAb, Human IgA1 (AM130)
Catalog No. 502054831
Encompass_Preferred
Antibody;1MG;Human IgA1/kappa;This product can recognize SARS-CoV-2 and SARS-CoV Spike Protein RBD domainn. No cross-reactivity is deteced with Spike Protein RBD domain of other coronaviruses, including MERS-CoV, HCoV-229E, HCoV-NL63, HCoV-OC43 and HCoV-HKU1.;Anti-SARS-CoV-2 Spike RBD Antibody, Chimeric mAb, Human IgA1 (AM130) is a chimeric monoclonal antibody combining the constant domains of the human IgA1 molecule with mouse variable regions.; The protein migrates as 27 kDa and 55 kDa under reducing (R) condition (SDS-PAGE).;Immobilized SARS-CoV-2 S protein RBD, His Tag (Cat. No.SPD-C52H1) at 2 μg/mL (100 μL/well) can bind Anti-SARS-CoV-2 Spike RBD Antibody, Chimeric mAb, Human IgA1 (Cat. No.S1N-M164) with a linear range of 0.24-3.9 ng/mL (QC tested).
Catalog No. 50-205-4831 Supplier ACROBiosystems Supplier No. S1NM1641MG
Antibody;1MG;Human IgA1/kappa;This product can recognize SARS-CoV-2 and SARS-CoV Spike Protein RBD domainn. No cross-reactivity is deteced with Spike Protein RBD domain of other coronaviruses, including MERS-CoV, HCoV-229E, HCoV-NL63, HCoV-OC43 and HCoV-HKU1.;Anti-SARS-CoV-2 Spike RBD Antibody, Chimeric mAb, Human IgA1 (AM130) is a chimeric monoclonal antibody combining the constant domains of the human IgA1 molecule with mouse variable regions.; The protein migrates as 27 kDa and 55 kDa under reducing (R) condition (SDS-PAGE).;Immobilized SARS-CoV-2 S protein RBD, His Tag (Cat. No.SPD-C52H1) at 2 μg/mL (100 μL/well) can bind Anti-SARS-CoV-2 Spike RBD Antibody, Chimeric mAb, Human IgA1 (Cat. No.S1N-M164) with a linear range of 0.24-3.9 ng/mL (QC tested).