Learn More
ACROBiosystems ACROBiosystems Anti-SARS-CoV-2 Spike RBD Neutralizing Antibody, Chimeric mAb, Cynomolgus IgG1 (AM122)
Catalog No. 502107032
Encompass_Preferred
Antibody;1MG;Human IgG1/kappa;This product is a specific antibody against SARS-CoV-2 Spike protein RBD domain. No cross-reactivity is detected with Spike protein RBD domain of other coronaviruses, including SARS-CoV, MERS-CoV, HCoV-229E, HCoV-NL63, HCoV-OC43 and HCoV-HKU1.;Anti-SARS-CoV-2 Spike RBD Neutralizing Antibody, Chimeric mAb, Cynomolgus IgG1 (AM122) (SPD-M201) is a chimeric monoclonal antibody recombinantly expressed from HEK293 cells, which combines the variable region of a mouse monoclonal antibody with Cynomolgus IgG1 constant domain. The mouse monoclonal antibody was obtained from a mouse immunized with recombinant SARS-CoV-2 Spike S1 Protein. ; The protein has a calculated MW of 48.8 kDa | 23.4 kDa. the protein migrates as 27 kDa and 55 kDa under reducing (R) condition, and 150 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized SARS-CoV-2 Spike RBD, His Tag (Cat. No. SPD-C52H1) at 2 μg/mL (100 μL/well) can bind Anti-SARS-CoV-2 Spike RBD Neutralizing Antibody
Catalog No. 50-210-7032 Supplier ACROBiosystems Supplier No. SPDM2011MG
Antibody;1MG;Human IgG1/kappa;This product is a specific antibody against SARS-CoV-2 Spike protein RBD domain. No cross-reactivity is detected with Spike protein RBD domain of other coronaviruses, including SARS-CoV, MERS-CoV, HCoV-229E, HCoV-NL63, HCoV-OC43 and HCoV-HKU1.;Anti-SARS-CoV-2 Spike RBD Neutralizing Antibody, Chimeric mAb, Cynomolgus IgG1 (AM122) (SPD-M201) is a chimeric monoclonal antibody recombinantly expressed from HEK293 cells, which combines the variable region of a mouse monoclonal antibody with Cynomolgus IgG1 constant domain. The mouse monoclonal antibody was obtained from a mouse immunized with recombinant SARS-CoV-2 Spike S1 Protein. ; The protein has a calculated MW of 48.8 kDa | 23.4 kDa. the protein migrates as 27 kDa and 55 kDa under reducing (R) condition, and 150 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized SARS-CoV-2 Spike RBD, His Tag (Cat. No. SPD-C52H1) at 2 μg/mL (100 μL/well) can bind Anti-SARS-CoV-2 Spike RBD Neutralizing Antibody