Learn More
ACROBiosystems Anti-SARS-CoV-2 Spike S2 protein Antibody, Human IgG4 (AS86)
Catalog No. 502054829
Encompass_Preferred
Antibody;1MG;Human IgG4/kappa;This product is a specific antibody against SARS-CoV-2 Spike S2 protein. Cross-reactivity with S2 protein of other coronaviruses has not been tested.;Anti-SARS-CoV-2 Spike S2 protein Antibody, Human IgG4 (AS86) is isolated from a SARS-CoV-2 infected patient and is recombinantly produced from CHO cells.; The protein migrates as 27 kDa and 55 kDa under reducing (R) condition (SDS-PAGE).;Immobilized SARS-CoV-2 S2 protein, His Tag (Cat. No. S2N-C52H5) at 2 μg/mL (100 μL/well) can bind Anti-SARS-CoV-2 Spike S2 protein Antibody, Human IgG4 (Cat. No. S2N-S86) with a linear range of 0.12-2 ng/mL (QC tested).
Catalog No. 50-205-4829 Supplier ACROBiosystems Supplier No. S2NS861MG
Antibody;1MG;Human IgG4/kappa;This product is a specific antibody against SARS-CoV-2 Spike S2 protein. Cross-reactivity with S2 protein of other coronaviruses has not been tested.;Anti-SARS-CoV-2 Spike S2 protein Antibody, Human IgG4 (AS86) is isolated from a SARS-CoV-2 infected patient and is recombinantly produced from CHO cells.; The protein migrates as 27 kDa and 55 kDa under reducing (R) condition (SDS-PAGE).;Immobilized SARS-CoV-2 S2 protein, His Tag (Cat. No. S2N-C52H5) at 2 μg/mL (100 μL/well) can bind Anti-SARS-CoV-2 Spike S2 protein Antibody, Human IgG4 (Cat. No. S2N-S86) with a linear range of 0.12-2 ng/mL (QC tested).