Learn More
ACROBiosystems ACROBiosystems Anti-SARS-CoV-2 Antibody IgG2 Titer Serologic Assay kit
Catalog No. 502107419
Encompass_Preferred
Kits;96TESTS;This assay kit employs a standard indirect-ELISA format, providing a rapid detection of Anti-SARS-CoV-2 antibodies in serum by SARS-CoV-2 Spike RBD. The Kit consists of Pre-coated with SARS-CoV-2 Spike RBD Microplate, an Anti-SARS-CoV-2 Antibody (Control, IgG2), an HRP-Anti-Human IgG2 secondary antibody.;Detection of Anti-SARS-CoV-2 Antibody, Human IgG2 titer by Indirect-ELISA Assay.
Immobilized SARS-CoV-2 Spike RBD at 2 μg/mL (100 μL/well) can bind Monoclonal Anti-SARS-CoV-2 Antibody, Human IgG2 in 1:50 human serum. Detection was performed using HRP-Conjugated Anti-human IgG2 antibody with sensitivity of 1000 ng/mL (QC tested).
Catalog No. 50-210-7419 Supplier ACROBiosystems Supplier No. RAST01596TESTS
Kits;96TESTS;This assay kit employs a standard indirect-ELISA format, providing a rapid detection of Anti-SARS-CoV-2 antibodies in serum by SARS-CoV-2 Spike RBD. The Kit consists of Pre-coated with SARS-CoV-2 Spike RBD Microplate, an Anti-SARS-CoV-2 Antibody (Control, IgG2), an HRP-Anti-Human IgG2 secondary antibody.;Detection of Anti-SARS-CoV-2 Antibody, Human IgG2 titer by Indirect-ELISA Assay.
Immobilized SARS-CoV-2 Spike RBD at 2 μg/mL (100 μL/well) can bind Monoclonal Anti-SARS-CoV-2 Antibody, Human IgG2 in 1:50 human serum. Detection was performed using HRP-Conjugated Anti-human IgG2 antibody with sensitivity of 1000 ng/mL (QC tested).