Learn More
Grainger Thread Sealant Tape, PTFE, 0.7 to 0.8sg, 1/2 in Width, 520 in Length, White Color
Catalog No. NC0776760
Encompass_Preferred
POLY-TEMP MD PTFE Tape is effective and simple to use. 99.9% pure, medium density, chemically inert tape that is suitable for oxygen service. UL Listed. Meets Mil. Spec. T-27730A. White color.
Catalog No. NC0776760 Supplier Grainger Supplier No. 4X543
POLY-TEMP MD PTFE Tape is effective and simple to use. 99.9% pure, medium density, chemically inert tape that is suitable for oxygen service. UL Listed. Meets Mil. Spec. T-27730A. White color.