Learn More
Vector Laboratories Anti-Sendai Virus HN Protein [1A6] Antibody | 100ug
SDP
Catalog No. NC1370067
Encompass
The Anti-Sendai Virus HN Protein [1A6] Antibody recognized the HN protein of Sendai virus. It is suitable for hemagglutination inhibition, neutralization and inmmunofluorescence applications.
Catalog No. NC1370067 Supplier Vector Laboratories Supplier No. EMS016
The Anti-Sendai Virus HN Protein [1A6] Antibody recognized the HN protein of Sendai virus. It is suitable for hemagglutination inhibition, neutralization and inmmunofluorescence applications.