Learn More
Immunology Consultants anti-SFTS Virus Antibody (Mouse - Clone #3G6) - Monoclonal
SDP
Catalog No. 5000435344
Encompass_Preferred
Clone 3G6 provides high‑specificity detection of SFTS virus, supporting ELISA, Western blotting, and antigen‑capture assay development. Its strong affinity and clean signal enable sensitive detection in serum, culture fluids, and purified antigen preparations. MSFTS‑55A‑3G6 is ideal for viral surveillance, diagnostic development, and studies examining SFTS pathogenesis. Its reproducible performance ensures dependable detection across repeated assays.
Catalog No. 50-004-35344 Supplier Immunology Consultants Supplier No. MSFTS55A3G6
Clone 3G6 provides high‑specificity detection of SFTS virus, supporting ELISA, Western blotting, and antigen‑capture assay development. Its strong affinity and clean signal enable sensitive detection in serum, culture fluids, and purified antigen preparations. MSFTS‑55A‑3G6 is ideal for viral surveillance, diagnostic development, and studies examining SFTS pathogenesis. Its reproducible performance ensures dependable detection across repeated assays.