Learn More
ECM Bioscience Shank1 (C-terminal region) Mouse Monoclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience SM3691
Encompass_Preferred
Clone M369 was generated from a sequence corresponding to amino acids in the C-terminal region of rat Shank1. This sequence has high homology to human and mouse Shank1. Accession #Q9WV48. This antibody detects a 225 kDa* protein corresponding to the molecular mass of Shank1 on SDS-PAGE immunoblots of adult mouse brain. Applications: WB, E; Species Reactivity: Hu, Rt, Ms; Size = 100 μl; Buffer = Mouse monoclonal, protein A purified antibody is supplied in 100µl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at -20°C. Stable for 1 year. Other Names = Spank1; Sstrip; Shank1a; Shank1; ProSAP
Catalog No. 50-206-8851