Learn More
Colorado Serum Co Anti Sheep Hemolysin, High Specificity For Sheep RBCs, Liquid Formulation, 10 ml
SDP
Supplier:  Colorado Serum Co 31162
Encompass
ANTI SHEEP HEMOLYSIN is a specialized laboratory reagent from COLORADO SERUM CO designed for immunology and serology applications. With high specificity for sheep red blood cells, it aids in agglutination and complement fixation assays. Its reliable rabbit-derived antibody formulation is provided in a ready-to-use 10 ml liquid format. This product is essential for researchers and diagnostic labs requiring precise and effective tools for complement pathway assessments.
Catalog No. NC0787194