Learn More
Seracare Life Sciences Inc ANTI-SHEEP IGG (G) AB FITC
Catalog No. 5066908
Encompass_Preferred
Affinity purified polyclonal antibody to Sheep IgG, heavy chains (gamma) made in Rabbit and labeled with fluorescein isothiocyanate (FITC). Product is in lyophilized form.
This item is not returnable. View return policy
Catalog No. 50-669-08 Supplier Seracare Life Sciences Inc Supplier No. 52300322
Affinity purified polyclonal antibody to Sheep IgG, heavy chains (gamma) made in Rabbit and labeled with fluorescein isothiocyanate (FITC). Product is in lyophilized form.