Learn More
Seracare Life Sciences Inc ANTI-SHEEP IGG (GAMMA) AB
Catalog No. 5066843
Encompass_Preferred
Affinity purified polyclonal antibody to Sheep IgG heavy chains (gamma) made in Rabbit. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
This item is not returnable. View return policy
Catalog No. 50-668-43 Supplier Seracare Life Sciences Inc Supplier No. 52100203
Affinity purified polyclonal antibody to Sheep IgG heavy chains (gamma) made in Rabbit. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.