Learn More
Seracare Life Sciences Inc ANTI-SHEEP IGG (H+L) AB HRP
Supplier:  Seracare Life Sciences Inc 52200372
Encompass_Preferred
Affinity purified polyclonal secondary antibody against Sheep IgG, heavy and light chains (whole IgG) made in Rabbit and labeled with horseradish peroxidase. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-672-34