Learn More
Seracare Life Sciences Inc ANTI-SHEEP IGG (H+L) ANTIBODY
Catalog No. 5066845
Encompass_Preferred
Affinity purified polyclonal antibody to Sheep IgG, both heavy and light chains (whole IgG) made in Rabbit. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
This item is not returnable. View return policy
Catalog No. 50-668-45 Supplier Seracare Life Sciences Inc Supplier No. 52100204
Affinity purified polyclonal antibody to Sheep IgG, both heavy and light chains (whole IgG) made in Rabbit. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.