Learn More
Rockland Immunochemicals Anti-SHEEP IgG (H&L) (DONKEY) Antibody DyLight™ 800 Conjugated (Min X Ch GP Ham Hs Hu Ms Rb & Rt Serum Proteins) - 613-745-168
SDP
Catalog No. 501058690
Encompass_Preferred
Anti-Sheep IgG (H&L) DyLight™800 Antibody generated in donkey detects reactivity to sheep IgG. No reaction was observed against Chicken, Guinea Pig, Hamster, Horse, Human, Mouse, Rabbit or Rat Serum Proteins.  This antibody will react with heavy chains of Sheep IgG and with light chains of most Sheep immunoglobulins. Secondary Antibodies are available in a variety of formats and conjugate types. When choosing a secondary antibody product, consideration must be given to species and immunoglobulin specificity, conjugate type, fragment and chain specificity, level of cross-reactivity, and host-species source and fragment composition. This Anti-Sheep IgG is conjugated to DyLight™800. This product is designed for immunofluorescence microscopy, fluorescence based plate assays (FLISA) and fluorescent western blotting. This product is also suitable for multiplex analysis, including multicolor imaging, utilizing various commercial platforms.
Catalog No. 50-105-8690 Supplier Rockland Immunochemicals Supplier No. 613745168
Anti-Sheep IgG (H&L) DyLight™800 Antibody generated in donkey detects reactivity to sheep IgG. No reaction was observed against Chicken, Guinea Pig, Hamster, Horse, Human, Mouse, Rabbit or Rat Serum Proteins.  This antibody will react with heavy chains of Sheep IgG and with light chains of most Sheep immunoglobulins. Secondary Antibodies are available in a variety of formats and conjugate types. When choosing a secondary antibody product, consideration must be given to species and immunoglobulin specificity, conjugate type, fragment and chain specificity, level of cross-reactivity, and host-species source and fragment composition. This Anti-Sheep IgG is conjugated to DyLight™800. This product is designed for immunofluorescence microscopy, fluorescence based plate assays (FLISA) and fluorescent western blotting. This product is also suitable for multiplex analysis, including multicolor imaging, utilizing various commercial platforms.