Learn More
Rockland Immunochemicals Sheep IgG (H&L) Antibody Peroxidase Conjugated 2 mg
SDP
Catalog No. 501943936
Encompass_Preferred
Anti-Sheep IgG antibody generated in rabbit detects specifically sheep IgG. This secondary antibody anti-Sheep is ideal for investigators who routinely perform ELISA Sandwich ELISA titration assays western-blot immunoprecipitation and more generally immunoassays.
Catalog No. 50-194-3936 Supplier Rockland Immunochemicals Supplier No. 6134302
Anti-Sheep IgG antibody generated in rabbit detects specifically sheep IgG. This secondary antibody anti-Sheep is ideal for investigators who routinely perform ELISA Sandwich ELISA titration assays western-blot immunoprecipitation and more generally immunoassays.