Learn More
Rockland Immunochemicals Sheep IgG (H&L) Antibody Peroxidase Conjugated 2 mg
SDP
Catalog No. 501943937
Encompass_Preferred
Anti-Sheep IgG antibody generated in donkey detects specifically sheep IgG. This secondary antibody anti-Sheep is ideal for investigators who routinely perform ELISA Sandwich ELISA titration assays western-blot immunoprecipitation and more generally immunoassays.
Catalog No. 50-194-3937 Supplier Rockland Immunochemicals Supplier No. 6137302
Anti-Sheep IgG antibody generated in donkey detects specifically sheep IgG. This secondary antibody anti-Sheep is ideal for investigators who routinely perform ELISA Sandwich ELISA titration assays western-blot immunoprecipitation and more generally immunoassays.