Learn More
Rockland Immunochemicals Anti-SHEEP IgG (H&L) (RABBIT) Antibody DyLight™ 680 Conjugated - 613-444-002
SDP
Supplier:  Rockland Immunochemicals 613444002
Encompass
Anti-Sheep IgG (H&L) DyLight™ 680 Antibody generated in rabbit detects reactivity to sheep IgG. This antibody will react with heavy chains of Sheep IgG and with light chains of most Sheep immunoglobulins. Secondary Antibodies are available in a variety of formats and conjugate types. When choosing a secondary antibody product, consideration must be given to species and immunoglobulin specificity, conjugate type, fragment and chain specificity, level of cross-reactivity, and host-species source and fragment composition. This Anti-Sheep IgG is conjugated to DyLight™680. This product is designed for immunofluorescence microscopy, fluorescence based plate assays (FLISA) and fluorescent western blotting. This product is also suitable for multiplex analysis, including multicolor imaging, utilizing various commercial platforms.
Catalog No. 50-105-8674