Learn More
Immunology Consultants Monoclonal Mouse anti-Sheep IgM Antibody Clone 2G3, 1.0 mg
SDP
Catalog No. 502414300
Encompass_Preferred
This mouse IgG1 is purified using Protein A chromatography. It is produced from tissue culture media obtained at our facility in The United States. Antibody concentration was determined using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody reacts with Sheep IgM as determined by ELISA and western blotting. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. 50-241-4300 Supplier Immunology Consultants Supplier No. MM35A2G3
This mouse IgG1 is purified using Protein A chromatography. It is produced from tissue culture media obtained at our facility in The United States. Antibody concentration was determined using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody reacts with Sheep IgM as determined by ELISA and western blotting. Optimal working dilutions should be determined experimentally by the investigator.