Learn More
Seracare Life Sciences Inc ANTI-SHEEPIGG(H+L)AB RESERVEAP
Catalog No. 5067245
Encompass_Preferred
Affinity purified polyclonal secondary antibody against Sheep IgG, heavy and light chains (whole IgG) made in Rabbit and labeled with alkaline phosphatase. ReserveAP conjugates represent the latest advances in KPL's conjugation chemistry and are more sensitive than KPL's standard AP conjugates. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
This item is not returnable. View return policy
Catalog No. 50-672-45 Supplier Seracare Life Sciences Inc Supplier No. 52200388
Affinity purified polyclonal secondary antibody against Sheep IgG, heavy and light chains (whole IgG) made in Rabbit and labeled with alkaline phosphatase. ReserveAP conjugates represent the latest advances in KPL's conjugation chemistry and are more sensitive than KPL's standard AP conjugates. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.