Learn More
STEMCELL Technologies Anti-SHIP (C-Terminus) Antibody, Polyclonal , Size: 100 µL
SDP
Supplier:  STEMCELL Technologies 60144
Encompass
SHIP (C-terminus) is critical for the function of SHIP protein. SH2-containing inositol phosphatase (SHIP) is a 145 kDa hematopoietic-restricted protein that becomes tyrosine-phosphorylated and associated with the adaptor protein, Shc, following cytokine, growth factor, chemokine, and immunoreceptor stimulation. SHIP also hydrolyzes the critical phosphatidylinositol (PI)-3-kinase (PI3K)-generated second messenger, PI-3,4,5-P3 (PIP3), to PI-3,4-P2 1,2 and therefore acts as an important negative regulator of the PI3K pathway. This antibody reacts with mouse and human full-length 145 kDa SHIP protein.
Catalog No. 50-197-5720