Learn More
ECM Bioscience SHP1 (C-terminal region) Mouse Monoclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience SM1601
Encompass_Preferred
Clone (M160) was generated from a recombinant protein containing amino acids in the C-terminal region of human SHP1. This sequence is highly conserved in rat and mouse SHP1. Accession #P29350. The antibody detects a 68 kDa* protein in human A431 and Jurkat cells, and does not cross-react with SHP2. Applications: WB, E, IP, ICC; Species Reactivity: Hu, Rt, Ms; Size = 100 μl; Buffer = Mouse monoclonal antibody purified with protein A chromatography is supplied in 100µl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = PTP1C, SHPTP1
Catalog No. 50-206-8842
Edge