Learn More
Selleck Chemical LLC Anti-Sialyl-Lewis A-1MG-25
SDP
Catalog No. 503136087
Encompass_Preferred
Anti-Sialyl-Lewis A is a human monoclonal antibody against the carbohydrate antigen sialyl-Lewis A (carbohydrate antigen 19-9 CA19-9) with potential antineoplastic activity MW 145 5 kD
Catalog No. 50-313-6087 Supplier Selleck Chemical LLC Supplier No. A23591MG 25PC
Add to Cart
Add to Cart
Request a Quote
Anti-Sialyl-Lewis A is a human monoclonal antibody against the carbohydrate antigen sialyl-Lewis A (carbohydrate antigen 19-9 CA19-9) with potential antineoplastic activity MW 145 5 kD