Learn More
ABclonal Technology SIGLEC8 Polyclonal Antibody, 100UL, Validated in:WB
SDP
Supplier:  ABclonal Technology A10514
Encompass_Preferred
Immunogen: Recombinant fusion protein containing a sequence corresponding to amino acids 17-363 of human Siglec-8 (NP_055257.2).•Host Species: Rabbit•Isotype: IgG•Species Reactivity:Human,Mouse,Rat•Validated Applications:WB•Swiss Prot: Q9NYZ4•Gene ID:27181•Purification Method:Affinity purification•Storage: Store at -20℃. Avoid freeze / thaw cycles.|Buffer: PBS with 0.02% sodium azide,50% glycerol,pH7.3.This product is available in 20UL, 50UL, 100UL, and 200UL. Please contact customer service for pricing and availability on alternative sizes.
Catalog No. 501310066
Edge