Learn More
Valutek™ Nanotek PE Coated Polypropylene Anti-Skid Boot Covers
Catalog No. 191502120
Change view
Click to view available options
Size:
Large
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-150-2120 X-Large
19-150-2119 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-150-2120 Supplier Valutek™ Supplier No. VTBTCVPPLVWXL
Add to Cart
Add to Cart
This item is not returnable. View return policy
Features a durable skid-resistant PVC sole design and offers protection from slips or falls as well as being impervious to liquids. Recommended for use in a cleanroom Class 1-10 (ISO 3-4) critical environments.
  • Clean, low-linting, and latex-free fabric
  • Non-skid PVC sole provides protection against wet and smooth surfaces
  • Coefficient of friction
Specifications
Size X-Large
Material Polypropylene
Latex Free Yes
Color White
Disposable Polyethylene
Sterility Non-sterile
Closure Type Elastic Top and Ankles
Sole Type Embossed PVC
Packaging Quantity 200/Cs.
Packaging Type 20/Bag, 10 Bags/Cs.
Product Type Controlled Environments Boot Covers
Cleanroom Class Class 1-10 (ISO 3-4)