Learn More
Valutek™ MacroTek Anti-Skid Polypropylene Shoe Covers
Catalog No. 191502108
Change view
Click to view available options
Size:
Medium
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-150-2108 X-Large
19-150-2106 Medium
19-150-0830 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-150-2108 Supplier Valutek™ Supplier No. VTSHCVASXL
Add to Cart
Add to Cart
This item is not returnable. View return policy
Manufactured with 100% non-linting, non-woven spunbond polypropylene fabric with stitched seams and sprayed latex anti-skid treads on the sole. Recommended for use in ISO 7 (Class 10,000) critical environments.
  • All Valutek™ shoe covers are tested and manufactured in ISO compliant facilities under individual inspection and strict process control to ensure Valutek™ quality standard and product specifications
  • Recommended for use in a cleanroom Class 10,000+ (ISO 7+) critical environment
  • Commonly used for protecting against dirt, grime, and certain dry particulates in nonhazardous environments
Specifications
Size X-Large
Material Polypropylene
Color Blue
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Ankle
Packaging Quantity 300/Cs.
Packaging Type 100/Bag, 3 Bags/Cs.
Product Type Controlled Environments Shoe Covers
Cleanroom Class Class 10,000 (ISO 7)