Learn More
Abcam ANTI-SODIUM POTASSIUM ATPASE, 100 μL, Rabbit recombinant monoclonal antibody, Clone EP1845Y, IgG, 0.548 mg/mL concentration, Human, Mouse, Rat, Chinese Hamster reactivity
Supplier:  Abcam AB76020100UL
Encompass_Preferred
This rabbit recombinant monoclonal antibody, clone EP1845Y, detects Sodium Potassium ATPase (ATP1A1), serving as a reliable plasma membrane loading control. It offers high specificity and lot-to-lot consistency, making it suitable for various scientific applications. The antibody shows broad species reactivity and has been extensively cited in peer-reviewed literature.
  • High specificity for the Sodium Potassium ATPase (ATP1A1) target ensures accurate detection.
  • Versatile application profile, including Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin), and Intracellular Flow Cytometry.
  • Functions effectively as a plasma membrane loading control in Western Blot experiments.
  • Recombinant monoclonal format guarantees superior batch-to-batch consistency and reproducibility.
  • Broad species reactivity (human, mouse, rat, Chinese hamster) supports diverse experimental designs.
Catalog No. NC0653903