Learn More
World Precision Instrument Anti-Solarization Fiber Optic Cables, 600 µm Core
SDP
Catalog No. 502538802
Encompass
Anti-Solarization Fibers stabilize to less than 5% degradation over a period of 20,000 hours after an initial transmission “burn-in” loss of less than 25%.
Catalog No. 50-253-8802 Supplier World Precision Instrument Supplier No. FO600ASSMA
Anti-Solarization Fibers stabilize to less than 5% degradation over a period of 20,000 hours after an initial transmission “burn-in” loss of less than 25%.