Learn More
Biolegend Purified (azide-free) anti-CRISPR (CAS9)
Supplier:  Biolegend 844301
Encompass_Preferred
Purified (azide-free) anti-CRISPR (CAS9) [7A9]; Isotype: Mouse IgG1, κ; Reactivity: S Pyogenes; Apps: ICC, WB, IP; Size: 100 ug
Catalog No. NC1040696