Learn More
Selleck Chemical LLC Anti-Sphingosine-1-phosphate
SDP
Catalog No. 503135271
Encompass_Preferred
Anti-Sphingosine-1-phosphate is a human IgG1 monoclonal anti-Sphingosine-1-phosphate antibody that targets lysosphingolipids thereby lowering the concentration of S1P in the extracellular fluid MW 145 74 KD
Catalog No. 50-313-5271 Supplier Selleck Chemical LLC Supplier No. A28571MG
Anti-Sphingosine-1-phosphate is a human IgG1 monoclonal anti-Sphingosine-1-phosphate antibody that targets lysosphingolipids thereby lowering the concentration of S1P in the extracellular fluid MW 145 74 KD