Learn More
Aviva Systems Biology Corp ST6GALNAC5 Antibody - middle region (ARP49986_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49986_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ST6GALNAC5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ST6GALNAC5. Alternate names: MGC3184, SIAT7E, ST6GalNAcVApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53612