Learn More
R&R Lotions ANTI-STAT. HAND LOTION 4GAL/CS
SDP
Catalog No. NC9139423
Encompass_Preferred
Gallon I.C. Antistatic Cleanroom Lotion is Fragrance and Dye Free and is enriched with Aloe Vera & Vitamins AD & E. The lotion relieves chapping, skin flaking, irritation and skin sensitivity caused by regular use of latex and nitrile gloves. This lotion helps to prevent Urticaria which is a simple
Catalog No. NC9139423 Supplier R&R Lotions Supplier No. ICLGALCR
Gallon I.C. Antistatic Cleanroom Lotion is Fragrance and Dye Free and is enriched with Aloe Vera & Vitamins AD & E. The lotion relieves chapping, skin flaking, irritation and skin sensitivity caused by regular use of latex and nitrile gloves. This lotion helps to prevent Urticaria which is a simple